Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
ENST00000366273.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013761.hg.1 |
TC06001153.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06013761.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
ENST00000366273.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010070.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168854.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168854.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168854.hg.1 |
ENST00000569097.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600168854.hg.1 |
NR_003288.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600168854.hg.1 |
TC0600010070.hg.1 |
Clariom D TC |