Assay Details
| Gene Symbol: SNORD116-29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003360.1 | 1 | 45 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000071.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4180 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16760341 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023729 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543875 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023730 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023728 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023734 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543888 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023736 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543883 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P30023731 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09543876 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16760341 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543956 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543947 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543847 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543844 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543838 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543872 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543884 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543863 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543882 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543953 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543828 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543851 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543840 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543831 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543869 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543888 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543842 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543846 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543950 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543961 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543958 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543829 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543877 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543886 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543818 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543874 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543880 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543949 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543951 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543945 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543854 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P09543875 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003360_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7982007 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7982006 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000679.hg.1 | ENST00000384516.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000679.hg.1 | NR_003360.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15000679.hg.1 | TC15000071.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500181223.hg.1 | ENST00000384516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500181223.hg.1 | NR_003360.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500181223.hg.1 | ENST00000384516.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500181223.hg.1 | NR_003360.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500181223.hg.1 | TC1500010700.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3310593 | NR_003360.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3310593 | NR_003360.1 | human_transcriptome |

SNORD116-29
Genomic Map