Assay Details
| Gene Symbol: PMS2P7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB017006.1 | - | 1 - 2 | 176 | 75 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | stromal antigen 3-like 3 (pseudogene) | STAG3L3 |
STAG3L1, STAG3L2, STAG3L3P |
-214.0 | Anti-sense | Chr. 7 - 72997295 - 73005943 on NCBI Build GRCh38 |
| Downstream |
speedy/RINGO cell cycle regulator family member E8, pseudogene |
SPDYE8P | 10.1kb | Anti-sense | Chr. 7 - 73019731 - 73029762 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013387.hg.1 | ENST00000363043.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700007961.hg.1 | ENST00000275546.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | ENST00000441237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013393.hg.1 | NR_028058.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011540.hg.1 | ENST00000447708.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000412694.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | ENST00000425039.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_003614.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700011543.hg.1 | NR_130940.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000333674.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013387.hg.1 | ENST00000363043.1 | Ensembl |

PMS2P7
Genomic Map