Assay Details
Gene Symbol: CXCR2P1 | |||||||
---|---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_002712.1 | 1 | 747 | 79 | |||
GenBank mRNA | BC033255.1 | - | LIT2061 IL8RBP |
1 | 747 | 79 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105373878 | LOC105373878 | -65.4kb | Anti-sense | Chr. 2 - 217978707 - 217992615 on NCBI Build GRCh38 | |
Downstream | C-X-C motif chemokine receptor 2 | CXCR2 |
CD182, CDw128b, CMKAR2, IL8R2, IL8RA, IL8RB |
64.0kb | Anti-sense | Chr. 2 - 218125290 - 218137253 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895816 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895814 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895815 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895805 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22020959 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22020957 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895808 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895810 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22020945 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895817 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P01895807 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P22020949 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 94989 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 847998 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 217141 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375859 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P38989 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 34756 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 61150 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P00597144 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_16_P00597144 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3375859 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3375859 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895825 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15383168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895817 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895798 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895823 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895805 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020961 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895804 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020945 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895797 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020959 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895814 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895807 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15383168 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895829 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895816 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020943 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895820 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895803 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895825 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020957 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895810 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895813 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895817 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P22020980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895801 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P01895798 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P38989 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P38989 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895833 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895789 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895799 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895842 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895803 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895820 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895836 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895829 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15383181 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895826 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895794 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895788 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895850 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895844 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895804 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895786 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895845 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895835 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895811 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895816 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895831 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895822 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895791 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895793 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895853 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895839 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895809 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895846 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895796 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895798 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895818 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895827 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895813 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P01895823 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00597144 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000443392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | ENST00000443392.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | NR_002712.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200015711.hg.1 | NR_002712.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200215289.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200215289.hg.1 | NR_002712.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200215289.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200215289.hg.1 | NR_002712.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200215289.hg.1 | TC0200015711.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000439871.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000443392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | ENST00000443392.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | NR_002712.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0200015711.hg.1 | NR_002712.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014994 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A014994 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1730621 | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1730621 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691507 | NR_002712.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1691507 | BC033255.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1730621 | - | - |
