Assay Details
| Gene Symbol: PRO0611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AI064823.1 | - | 1 | 247 | 146 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AF111851.1 | - | 247 | Chr. - - [-] on Build | 146 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 103C |
SNORD103C |
HBII-251, SNORD85 |
-19.8kb | Anti-sense | Chr. 1 - 30968163 - 30968237 on NCBI Build GRCh38 |
| Downstream | Na+/K+ transporting ATPase interacting 1 | NKAIN1 | FAM77C | 191.1kb | Anti-sense | Chr. 1 - 31179270 - 31239887 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3232363 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3861 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3862 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 43257 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3232363 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3232363 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_19_P00325583 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117411 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117393 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140048 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140033 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140053 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117411 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140049 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140020 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117368 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140059 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117395 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140040 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140006 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140057 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140012 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140034 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117375 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140055 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140044 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140019 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140027 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140030 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140022 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045263 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139999 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117397 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140045 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140052 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20139997 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140003 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140026 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140033 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15045262 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140028 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117398 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140016 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140029 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117369 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140046 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140031 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00117393 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140043 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140008 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P20140048 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117355 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117383 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117363 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117368 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117378 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117410 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117351 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117380 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117416 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117360 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117413 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117359 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117397 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15045267 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117357 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117403 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15045262 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117354 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117393 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117377 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P15045263 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117371 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117396 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117412 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117374 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117389 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117407 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117375 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117387 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117399 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117385 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P00117401 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.1 ST Array | 7899602 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2230374 | AI064823.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2230374 | NR_002762.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2230374 | AI064823.1 | human_transcriptome |

PRO0611
Genomic Map