Assay Details
| Gene Symbol: SNORA71A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002911.1 | 1 | 67 | 94 | ||
| GenBank mRNA | Y11165.1 | - | 1 | 67 | 94 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BM918074.1 | - | 69 | Chr. - - [-] on Build | 94 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small nucleolar RNA host gene 17 | SNHG17 | Sense | Chr. 20 - 38420592 - 38435375 on NCBI Build GRCh38 | ||
| Downstream |
small nucleolar RNA, H/ACA box 71C |
SNORA71C | U71c | 2.2kb | Sense | Chr. 20 - 38429667 - 38429804 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000839.hg.1
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3916 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156466 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156448 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156459 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156457 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804518 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156455 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804531 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156461 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156449 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156448 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156462 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156453 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P31804517 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P11156466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156461 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156499 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156454 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156480 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156436 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156503 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156449 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156464 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17190968 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156494 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156452 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156510 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156456 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156488 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17190966 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156493 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156466 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156458 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156512 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156459 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156471 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156507 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156502 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17190965 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156490 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P11156501 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002911_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8066259 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8066258 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1565858_PM_at | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1565859_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11763920_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11763920_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011551.hg.1 | ENST00000364729.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011551.hg.1 | ENST00000439912.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011551.hg.1 | NR_002911.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20011551.hg.1 | TC20000839.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2000163594.hg.1 | ENST00000364729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163594.hg.1 | NR_002911.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000163594.hg.1 | TC2000009070.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000163594.hg.1 | ENST00000364729.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000163594.hg.1 | NR_002911.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1565859_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565858_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565858_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1565859_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565858_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1565859_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3245605 | NR_002911.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3245605 | Y11165.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245605 | NR_002911.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3245605 | Y11165.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245605 | NR_002911.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3245605 | Y11165.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245605 | NR_002911.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3245605 | Y11165.1 | human_transcriptome |

SNORA71A
Genomic Map