Assay Details
| Gene Symbol: SNORD116@ | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF241255.1 | - | LIT1354 PWCR1 |
1 | 847 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
small nucleolar RNA, C/D box 116-21 |
SNORD116-21 | HBII-85-21 | Sense | Chr. 15 - 25088803 - 25088896 on NCBI Build GRCh38 | |
| Downstream |
small nucleolar RNA, C/D box 116-22 |
SNORD116-22 | HBII-85-22 | 1.0kb | Sense | Chr. 15 - 25089922 - 25090015 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SNORD116@
Genomic Map