Assay Details
| Gene Symbol: OAT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000274.3 | NP_000265.1 | 5 - 6 | 781 | 80 | |
| NM_001171814.1 | NP_001165285.1 | 4 - 5 | 553 | 80 | ||
| NM_001322965.1 | NP_001309894.1 | 5 - 6 | 776 | 80 | ||
| NM_001322966.1 | NP_001309895.1 | 6 - 7 | 1096 | 80 | ||
| NM_001322967.1 | NP_001309896.1 | 6 - 7 | 901 | 80 | ||
| NM_001322968.1 | NP_001309897.1 | 7 - 8 | 988 | 80 | ||
| NM_001322969.1 | NP_001309898.1 | 6 - 7 | 868 | 80 | ||
| NM_001322970.1 | NP_001309899.1 | 7 - 8 | 1084 | 80 | ||
| NM_001322971.1 | NP_001309900.1 | 3 - 4 | 460 | 80 | ||
| NM_001322974.1 | NP_001309903.1 | 5 - 6 | 667 | 80 | ||
| XM_017016279.1 | XP_016871768.1 | 5 - 6 | 2576 | 80 | ||
| GenBank mRNA | AB208817.1 | - | 5 - 6 | 2566 | 80 | |
| AK296032.1 | - | 4 - 5 | 500 | 80 | ||
| AK312561.1 | - | 5 - 6 | 728 | 80 | ||
| AK315947.1 | - | 4 - 5 | 500 | 80 | ||
| BC000964.2 | - | 5 - 6 | 709 | 3449883 | 80 | |
| BC016928.1 | - | 5 - 6 | 713 | 3913352 | 80 | |
| CD683561.1 | - | 3 - 4 | 429 | 80 | ||
| CR457045.1 | - | 4 - 5 | 648 | 80 | ||
| CR749808.1 | - | 4 - 5 | 503 | 80 | ||
| M12267.1 | - | 5 - 6 | 702 | 80 | ||
| M14963.1 | - | 4 - 5 | 692 | 80 | ||
| M23204.1 | - | 5 - 6 | 699 | 80 | ||
| Y07511.1 | - | 4 - 5 | 688 | 80 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

OAT
Genomic Map