Assay Details
| Gene Symbol: BCL2A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114735.1 | NP_001108207.1 | 2 - 3 | 652 | 84 | |
| GenBank mRNA | CD640106.1 | - | 2 - 3 | 418 | 84 | |
More Information
Set Membership: |
|

BCL2A1
Genomic Map