Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | small integral membrane protein 13 | SMIM13 | C6orf228 | -53.7kb | Anti-sense | Chr. 6 - 11094033 - 11138738 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC105374928 | LOC105374928 | 211.7kb | Anti-sense | Chr. 6 - 11417401 - 11526218 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map