Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 4462 | MIR4462 | -95.8kb | Sense | Chr. 6 - 37555365 - 37555422 on NCBI Build GRCh38 | |
| Downstream | zinc finger AN1-type containing 3 | ZFAND3 | TEX27 | 167.2kb | Anti-sense | Chr. 6 - 37819496 - 38154624 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map