Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | GTF2I repeat domain containing 1 | GTF2IRD1 |
BEN, CREAM1, GTF3, MUSTRD1, RBAP2, WBS, WBSCR11, WBSCR12, hMusTRD1alpha1 |
-31.0kb | Sense | Chr. 7 - 74453790 - 74603070 on NCBI Build GRCh38 |
| Downstream | general transcription factor IIi | GTF2I |
BAP135, BTKAP1, DIWS, GTFII-I, IB291, SPIN, TFII-I, WBS, WBSCR6 |
23.8kb | Sense | Chr. 7 - 74657665 - 74760692 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map