Assay Details
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | kallikrein related peptidase 5 | KLK5 |
KLK-L2, KLKL2, SCTE |
-9.1kb | Sense | Chr. 19 - 50943299 - 50953088 on NCBI Build GRCh38 |
Downstream | kallikrein related peptidase 7 | KLK7 |
PRSS6, SCCE, hK7 |
13.7kb | Sense | Chr. 19 - 50976479 - 50984064 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
