Assay Details
| Gene Symbol: NFATC2 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927566.1 | - | LIT3567 AY927566 |
1 | 255 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3194 | MIR3194 | mir-3194 | -22.3kb | Sense | Chr. 20 - 51452905 - 51452977 on NCBI Build GRCh38 |
| Downstream | ATPase phospholipid transporting 9A (putative) | ATP9A | ATPIIA | 121.1kb | Sense | Chr. 20 - 51596775 - 51768411 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

NFATC2
Genomic Map