Assay Details
| Gene Symbol: STYXL1 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY927581.1 | - | LIT3580 AY927581 |
7 - 8 | 749 | 89 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | transmembrane protein 120A | TMEM120A |
NET29, TMPIT |
-5.7kb | Sense | Chr. 7 - 75986837 - 75994674 on NCBI Build GRCh38 |
| Downstream | malate dehydrogenase 2 | MDH2 |
M-MDH, MDH, MGC:3559, MOR1 |
47.1kb | Anti-sense | Chr. 7 - 76048019 - 76067508 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

STYXL1
Genomic Map