Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 1510 | LINC01510 | -15.3kb | Sense | Chr. 7 - 116563594 - 116614820 on NCBI Build GRCh38 | |
| Downstream |
MET proto-oncogene, receptor tyrosine kinase |
MET |
AUTS9, DFNB97, HGFR, RCCP2, c-Met |
41.4kb | Anti-sense | Chr. 7 - 116672359 - 116798386 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map