Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
DEF6, guanine nucleotide exchange factor |
DEF6 |
IBP, SLAT, SWAP70L |
-100.0kb | Anti-sense | Chr. 6 - 35297818 - 35321771 on NCBI Build GRCh38 |
| Downstream | Fanconi anemia complementation group E | FANCE |
FACE, FAE |
29.6kb | Anti-sense | Chr. 6 - 35452339 - 35467106 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map