Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | TATA-box binding protein associated factor 9b | TAF9B |
DN-7, DN7, TAF9L, TAFII31L, TFIID-31 |
-63.2kb | Anti-sense | Chr. X - 78129748 - 78139682 on NCBI Build GRCh38 |
| Downstream | cysteinyl leukotriene receptor 1 | CYSLTR1 |
CYSLT1, CYSLT1R, CYSLTR, HMTMF81 |
67.9kb | Anti-sense | Chr. X - 78271468 - 78327691 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Genomic Map