Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | nuclear receptor interacting protein 1 | NRIP1 | RIP140 | -428.0kb | Sense | Chr. 21 - 14961235 - 15065903 on NCBI Build GRCh38 |
| Downstream | ubiquitin specific peptidase 25 | USP25 | USP21 | 233.8kb | Anti-sense | Chr. 21 - 15729947 - 15880072 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map