Assay Details
| Gene Symbol: LOC101928911 | |||||||
|---|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | RNADB ID | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_110869.1 | 2 - 3 | 262 | 105 | |||
| GenBank mRNA | AY927641.1 | - | LIT3622 AY927641 |
2 - 3 | 263 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | akirin 2 | AKIRIN2 |
C6orf166, FBI1, dJ486L4.2 |
-82.8kb | Anti-sense | Chr. 6 - 87674860 - 87702267 on NCBI Build GRCh38 |
| Downstream | sperm acrosome associated 1 | SPACA1 | SAMP32 | 137.1kb | Sense | Chr. 6 - 88046907 - 88066832 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LOC101928911
Genomic Map