Assay Details
| Gene Symbol: RMST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024037.1 | 6 - 7 | 676 | 156 | ||
| GenBank mRNA | AK056164.1 | - | 6 - 7 | 680 | 156 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1251 | MIR1251 |
MIRN1251, hsa-mir-1251 |
-2.7kb | Sense | Chr. 12 - 97491909 - 97491978 on NCBI Build GRCh38 |
| Downstream | microRNA 135a-2 | MIR135A2 |
MIRN135-2, MIRN135A2, mir-135a-2 |
67.8kb | Sense | Chr. 12 - 97563812 - 97563911 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12000753.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000610287.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000611651.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000611980.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000613072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000616309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000616886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000617263.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000619945.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | ENST00000621935.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009556.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12009556.hg.1 | TC12000753.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200162783.hg.1 | TC1200008551.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000538559.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000541282.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000547996.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000548886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000552411.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000610287.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000611651.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000611980.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000613072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000616309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000616886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000617263.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000618704.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000619945.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | ENST00000621935.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008551.hg.1 | NR_024037.1 | Refseq |

RMST
Genomic Map