Assay Details
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | SAM and SH3 domain containing 3 | SASH3 |
753P9, CXorf9, HACS2, SH3D6C, SLY |
-33.8kb | Sense | Chr. X - 129779916 - 129795201 on NCBI Build GRCh38 |
| Downstream | UTP14A small subunit processome component | UTP14A |
NYCO16, SDCCAG16, dJ537K23.3 |
77.0kb | Sense | Chr. X - 129906121 - 129929762 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

Genomic Map