Assay Details
| Gene Symbol: NPHP3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002811.1 | 2 - 3 | 128 | 76 | ||
| GenBank mRNA | BC036463.1 | - | 3 - 4 | 791 | 76 | |
| BC039375.1 | - | 2 - 3 | 128 | 76 | ||
| BC048124.1 | - | 2 - 3 | 127 | 76 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000489343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | ENST00000504440.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008850.hg.1 | NR_002811.1 | Refseq |

NPHP3-AS1
Genomic Map