Assay Details
Gene Symbol: SNORD127 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_003691.1 | 1 | 56 | 84 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | uncharacterized LOC105370474 | LOC105370474 | -26.9kb | Anti-sense | Chr. 14 - 45082427 - 45083991 on NCBI Build GRCh38 | |
Downstream | FK506 binding protein 3 | FKBP3 |
FKBP-25, FKBP-3, FKBP25, PPIase |
4.6kb | Anti-sense | Chr. 14 - 45115599 - 45134529 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC14000265.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_4250 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720253 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16699153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720250 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720261 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261910 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720282 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16699159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720255 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720245 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720244 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720248 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16699159 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720267 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720281 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720266 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720244 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720249 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720272 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720263 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720270 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720243 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720276 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720250 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720261 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720274 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720246 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261910 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720260 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261923 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720245 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720252 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720247 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261880 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720262 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261882 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720239 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720255 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720236 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720279 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720280 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720233 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720256 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720235 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16699153 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720253 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261920 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P09261877 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P29720282 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261905 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261913 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261884 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16699152 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261876 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261920 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261878 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261887 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261873 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261901 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16699156 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16699148 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261910 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261922 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16699158 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261892 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261893 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261917 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261897 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261906 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261908 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16699149 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261915 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261881 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261907 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261909 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261903 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09261899 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR14002856.hg.1 | ENST00000458892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR14002856.hg.1 | NR_003691.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR14002856.hg.1 | TC14000265.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1400007038.hg.1 | ENST00000458892.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400007038.hg.1 | NR_003691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400148905.hg.1 | ENST00000458892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400148905.hg.1 | NR_003691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400148905.hg.1 | ENST00000458892.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400148905.hg.1 | NR_003691.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400148905.hg.1 | TC1400007038.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1400007038.hg.1 | ENST00000458892.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400007038.hg.1 | NR_003691.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3242633 | NR_003691.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_3242633 | NR_003691.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3242633 | NR_003691.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3242633 | NR_003691.1 | human_transcriptome |
