Assay Details
| Gene Symbol: E2F6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278276.1 | NP_001265205.1 | 1 - 2 | 405 | 102 | |
| GenBank mRNA | AY551348.1 | - | 1 - 2 | 107 | 102 | |
| BU076470.1 | - | 1 - 2 | 83 | 102 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | long intergenic non-protein coding RNA 570 | LINC00570 | ncRNA-a5 | -54.1kb | Anti-sense | Chr. 2 - 11393981 - 11403077 on NCBI Build GRCh38 |
| Downstream | growth regulation by estrogen in breast cancer 1 | GREB1 | 67.9kb | Anti-sense | Chr. 2 - 11534116 - 11642788 on NCBI Build GRCh38 |
More Information
Additional Information:
This assay is in the 5'UTR region of the AY551348.1, NM_001278276.1 transcripts.
Set Membership: |
|

E2F6
Genomic Map