Assay Details
| Gene Symbol: POT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003104.1 | 14 - 15 | 1973 | 160 | ||
| GenBank mRNA | AY823524.1 | - | 10 - 11 | 1383 | 161 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | chromosome 7 open reading frame 77 | C7orf77 | -46.3kb | Sense | Chr. 7 - 124777292 - 124790810 on NCBI Build GRCh38 | |
| Downstream | POT1 antisense RNA 1 | POT1-AS1 | 88.7kb | Anti-sense | Chr. 7 - 124929873 - 125145234 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001830.hg.1
Set Membership: |
|

POT1
Genomic Map