| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
TC05000442.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
ENST00000296619.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
ENST00000399043.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
ENST00000405460.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
NM_032119.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05006265.hg.1 |
NR_003149.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
ENST00000405460.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
NM_032119.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
NR_003149.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
ENST00000405460.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
NM_032119.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
NR_003149.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500155193.hg.1 |
TC0500008055.hg.1 |
Clariom D TC |