Assay Details
| Gene Symbol: SNX29P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BX648280.1 | - | 2 - 3 | 672 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC101928188 | LOC101928188 | -70.5kb | Anti-sense | Chr. 16 - 29210768 - 29221006 on NCBI Build GRCh38 | |
| Downstream | nuclear pore complex interacting protein family member B11 | NPIPB11 | NPIP | 78.8kb | Anti-sense | Chr. 16 - 29381318 - 29403802 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000356328.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000398878.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000502626.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000507381.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000509141.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000569730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000603801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | ENST00000604430.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600007392.hg.1 | NR_002939.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600008812.hg.1 | ENST00000563190.1 | Ensembl |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 119017 | - | - |

SNX29P2
Genomic Map