| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023832.hg.1 |
TC07001577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023832.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023832.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023834.hg.1 |
TC07001577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023834.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023834.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023835.hg.1 |
TC07001577.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023835.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07023835.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000616845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
ENST00000616845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183516.hg.1 |
TC0700011703.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000421293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000616845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000421293.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000432193.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
ENST00000616845.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183517.hg.1 |
TC0700011703.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000542586.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000542586.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183513.hg.1 |
TC0700011703.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000359941.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000416560.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000542586.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
ENST00000610086.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
NR_015381.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700183515.hg.1 |
TC0700011703.hg.1 |
Clariom D TC |