Assay Details
| Gene Symbol: SIGLEC16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC039008.1 | - | 10 | 1813 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | sialic acid binding Ig like lectin 11 | SIGLEC11 | -13.6kb | Anti-sense | Chr. 19 - 49948993 - 49961172 on NCBI Build GRCh38 | |
| Downstream | vaccinia related kinase 3 | VRK3 | 653.0 | Anti-sense | Chr. 19 - 49976467 - 50025954 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human CGH Microarray 4x180K | 60145 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 933709 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 92898 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306024 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3230948 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3901 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_3902 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 8510 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 2972 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3306024 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3230948 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3230948 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3306024 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988329 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988317 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626360 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988331 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626351 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988313 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153408 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626357 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988320 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988332 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988325 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626359 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988333 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988326 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988321 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988330 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988314 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988316 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153367 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626362 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988318 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988328 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626358 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153407 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153377 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988319 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988327 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988312 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626354 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988323 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988322 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626361 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626365 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988324 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10988312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153377 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153407 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988328 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988330 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626351 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988314 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153377 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626358 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988319 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988321 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988323 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626362 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988318 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988331 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626359 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988312 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988329 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988325 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153367 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626361 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153408 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626357 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988322 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626354 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988333 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17153366 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988320 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988326 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988332 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626365 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988317 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31626360 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988313 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988316 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988324 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10988327 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153366 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10988312 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17153377 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P03460129 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P41218537 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P13056158 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03460129 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P942709 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988325 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988317 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17153366 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988322 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17153367 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988321 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988314 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988331 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988326 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988328 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10988320 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P17153408 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03460129 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_002825_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030567 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030564 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030566 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030565 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030568 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8030563 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1561880_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11762044_PM_a_at | - | - |
| Affymetrix Human Genome U219 Array | 11762044_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561880_PM_a_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1561880_a_at | - | - |
| Affymetrix GeneChip Human Genome U133 Plus 2.0 Array based on a custom CDF (REFSEQ_7) | NR_002825_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1561880_a_at7 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007108 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007108 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2229261 | NR_002825.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2229261 | BC030222.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2229261 | NR_002825.1 | human_transcriptome |

SIGLEC16
Genomic Map