Assay Details
Gene Symbol: ZNF138 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073389.1 | 3 | 149 | |||
GenBank mRNA | AL834121.1 | - | 1 | 1704 | 149 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | zinc finger protein 107 | ZNF107 |
Y8, ZFD25, ZNF588, smap-7 |
-140.2kb | Sense | Chr. 7 - 64665012 - 64711582 on NCBI Build GRCh38 |
Downstream | eukaryotic translation elongation factor 1 delta pseudogene 4 | EEF1DP4 | 10.7kb | Sense | Chr. 7 - 64862951 - 64864450 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
