Assay Details
| Gene Symbol: SNORA75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002921.1 | 1 | 28 | 64 | ||
| GenBank mRNA | AJ007015.1 | - | 1 | 28 | 64 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 7 | B3GNT7 | beta3GnT7 | -47.0kb | Anti-sense | Chr. 2 - 231395624 - 231401164 on NCBI Build GRCh38 |
| Downstream |
small nucleolar RNA, C/D box 20 |
SNORD20 |
RNU20, U20 |
508.0 | Sense | Chr. 2 - 231456444 - 231456523 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002860.hg.1
Set Membership: |
|

SNORA75
Genomic Map