Assay Details
| Gene Symbol: DMKN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011527501.2 | XP_011525803.1 | 11 - 12 | 1324 | 118 | |
| XM_011527503.2 | XP_011525805.1 | 10 - 11 | 1288 | 118 | ||
| XM_017027475.1 | XP_016882964.1 | 4 - 5 | 279 | 118 | ||
| GenBank mRNA | EF158326.1 | - | 5 - 6 | 341 | 118 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the EF158326.1 transcripts.
Set Membership: |
|

DMKN
Genomic Map