Assay Details
| Gene Symbol: SPRYD7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023351.2 | 1 - 2 | 366 | 101 | ||
| GenBank mRNA | DA388141.1 | - | 1 - 2 | 191 | 101 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
CTAGE family member 10, pseudogene |
CTAGE10P | -28.1kb | Anti-sense | Chr. 13 - 49890409 - 49893380 on NCBI Build GRCh38 | |
| Downstream | deleted in lymphocytic leukemia 2 (non-protein coding) | DLEU2 |
1B4, BCMSUN, DLB2, LEU2, LINC00022, MIR15AHG, NCRNA00022, RFP2OS, TRIM13OS |
46.1kb | Sense | Chr. 13 - 49982552 - 50125541 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

SPRYD7
Genomic Map