Assay Details
| Gene Symbol: DTX2P1-UPK3BP1-PMS2P11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023383.1 | 6 - 7 | 1094 | 98 | ||
| GenBank mRNA | DR007973.1 | - | 1 - 2 | 258 | 98 | |
| Gene Symbol: LOC105375355 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005250613.4 | XP_005250670.2 | 3 - 4 | 768 | 98 | |
| XM_017012885.1 | XP_016868374.1 | 3 - 4 | 768 | 98 | ||
| XM_017012886.1 | XP_016868375.1 | 3 - 4 | 768 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | deltex E3 ubiquitin ligase 2 | DTX2 | RNF58 | -5.7kb | Sense | Chr. 7 - 76461655 - 76505995 on NCBI Build GRCh38 |
| Downstream | speedy/RINGO cell cycle regulator family member E17 | SPDYE17 | 11.7kb | Anti-sense | Chr. 7 - 77023682 - 77031104 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07003311.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700008097.hg.1 | ENST00000513375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008097.hg.1 | ENST00000513375.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006900.hg.1 | TC07003311.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07006900.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197457.hg.1 | TC0700013392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000421583.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000425797.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000428678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013392.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000451831.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | ENST00000584900.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | NR_023383.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700197458.hg.1 | TC0700013392.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008097.hg.1 | ENST00000513375.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008097.hg.1 | ENST00000513375.1 | Ensembl |

DTX2P1-UPK3BP1-PMS2P11
Genomic Map