Assay Details
| Gene Symbol: LINC00470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_023927.1 | 2 - 3 | 282 | 95 | ||
| GenBank mRNA | AF295730.1 | - | 2 - 3 | 304 | 95 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC388456 | LOC388456 | -344.3kb | Sense | Chr. 18 - 926083 - 928129 on NCBI Build GRCh38 | |
| Downstream | uncharacterized LOC101926997 | LOC101926997 | 1.2mb | Sense | Chr. 18 - 2517250 - 2519260 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
Set Membership: |
|

LINC00470
Genomic Map