Assay Details
| Gene Symbol: LINC00158 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024027.2 | 1 - 2 | 523 | 67 | ||
| GenBank mRNA | AY035382.1 | - | 1 - 2 | 522 | 67 | |
| AY035383.1 | - | 1 - 2 | 522 | 67 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC339622 | LOC339622 | -372.0kb | Anti-sense | Chr. 21 - 24840550 - 25057746 on NCBI Build GRCh38 | |
| Downstream | MIR155 host gene | MIR155HG |
BIC, MIRHG2, NCRNA00172 |
130.4kb | Anti-sense | Chr. 21 - 25562145 - 25575168 on NCBI Build GRCh38 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000332.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003777.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003777.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003777.hg.1 | TC21000332.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003778.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003778.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003778.hg.1 | TC21000332.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | TC2100007784.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152504.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | TC2100007784.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152507.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000332587.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000439840.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000440205.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | ENST00000441013.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007784.hg.1 | NR_024027.2 | Refseq |

LINC00158
Genomic Map