Assay Details
| Gene Symbol: ADGRE4P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024075.1 | 8 - 9 | 879 | 92 | ||
| GenBank mRNA | AF489700.1 | - | 8 - 9 | 879 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | adhesion G protein-coupled receptor E1 | ADGRE1 |
EMR1, TM7LN3 |
-31.8kb | Anti-sense | Chr. 19 - 6887549 - 6940453 on NCBI Build GRCh38 |
| Downstream | microtubule affinity regulating kinase 1 pseudogene | FLJ25758 | 30.3kb | Sense | Chr. 19 - 7004953 - 7006415 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

ADGRE4P
Genomic Map