Assay Details
| Gene Symbol: LINC00521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024182.1 | 6 - 7 | 1146 | 87 | ||
| GenBank mRNA | BC031252.1 | - | 6 - 7 | 1146 | 87 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ankyrin repeat and SOCS box containing 2 | ASB2 | ASB-2 | -24.3kb | Anti-sense | Chr. 14 - 93934153 - 93976796 on NCBI Build GRCh38 |
| Downstream |
OTU deubiquitinase, ubiquitin aldehyde binding 2 |
OTUB2 |
C14orf137, OTB2, OTU2 |
12.7kb | Sense | Chr. 14 - 94016083 - 94048933 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

LINC00521
Genomic Map