Assay Details
| Gene Symbol: ESCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF306677.1 | - | 1 | 1241 | 104 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | coiled-coil domain containing 25 | CCDC25 | -1.7kb | Anti-sense | Chr. 8 - 27733316 - 27772656 on NCBI Build GRCh38 | |
| Downstream | PDZ binding kinase | PBK |
CT84, HEL164, Nori-3, SPK, TOPK |
33.1kb | Anti-sense | Chr. 8 - 27809621 - 27838095 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ESCO2
Genomic Map