Assay Details
| Gene Symbol: LOC283713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK094982.1 | - | 1 | 949 | 113 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | Kruppel like factor 13 | KLF13 |
BTEB3, FKLF2, NSLP1, RFLAT-1, RFLAT1 |
-39.7kb | Anti-sense | Chr. 15 - 31326855 - 31435665 on NCBI Build GRCh38 |
| Downstream | OTU deubiquitinase 7A | OTUD7A |
C15orf16, C16ORF15, CEZANNE2, OTUD7 |
3.2kb | Sense | Chr. 15 - 31483126 - 31655339 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565159 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565163 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565165 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565161 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P30051926 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P09565158 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 396736 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 363242 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 321632 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 228261 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 407644 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 377465 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3439713 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53839 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3439713 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P40348492 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P40348492 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix HT HG-U133+ PM Array Plate | 230783_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000307050.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000558371.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560536.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | ENST00000560598.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010845.hg.1 | NM_130901.2 | Refseq |
| Human Genome U133 Plus 2.0 Array | 230783_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 230783_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 230783_at8 | - | - |
| Human Genome U95 Set | 51770_PM_at | - | - |
| Human Genome U95 Set | 51770_at | - | - |
| Human Genome U133 Set | 230783_at | - | - |
| Human Genome U133 Set | 230783_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 182460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1911213 | AK094982.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1911213 | AK094982.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1911213 | AK094982.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | ILMN_70827 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_70827 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1911213 | AK094982.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1911213 | AK094982.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1911213 | AK094982.1 | human_transcriptome |

LOC283713
Genomic Map