Assay Details
| Gene Symbol: LOC100129115 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK095213.1 | - | 1 | 1448 | 67 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 6784 | MIR6784 | hsa-mir-6784 | -12.2kb | Sense | Chr. 17 - 45114367 - 45114433 on NCBI Build GRCh38 |
| Downstream | acyl-CoA binding domain containing 4 | ACBD4 | HMFT0700 | 4.0kb | Anti-sense | Chr. 17 - 45132600 - 45144181 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100129115
Genomic Map