Assay Details
| Gene Symbol: LOC728147 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096262.1 | - | 1 | 2144 | 106 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | uncharacterized LOC283440 | LOC283440 | -20.9kb | Sense | Chr. 12 - 2761200 - 2771669 on NCBI Build GRCh38 | |
| Downstream | FK506 binding protein 4 | FKBP4 |
FKBP51, FKBP52, FKBP59, HBI, Hsp56, PPIase, p52 |
64.0 | Anti-sense | Chr. 12 - 2794942 - 2805423 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC728147
Genomic Map