Assay Details
| Gene Symbol: LOC100131170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096606.1 | - | 1 | 1054 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | BAIAP2 antisense RNA 1 (head to head) | BAIAP2-AS1 | -75.8kb | Sense | Chr. 17 - 81029133 - 81034719 on NCBI Build GRCh38 | |
| Downstream | apoptosis associated tyrosine kinase | AATK |
AATYK, AATYK1, LMR1, LMTK1, PPP1R77, p35BP |
4.2kb | Sense | Chr. 17 - 81117295 - 81166072 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC100131170
Genomic Map