Assay Details
Gene Symbol: LOC642335 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK098072.1 | - | 1 | 249 | 82 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | DB195384.1 | - | 1 | 249 | Chr. 6 - 538127 - 538632 [-] on Build GRCh38 | 82 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | exocyst complex component 2 | EXOC2 |
SEC5, SEC5L1, Sec5p |
Sense | Chr. 6 - 485138 - 693141 on NCBI Build GRCh38 | |
Downstream | HUS1 checkpoint clamp component B | HUS1B | 117.3kb | Sense | Chr. 6 - 655939 - 656964 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
