Assay Details
| Gene Symbol: TMCO5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098530.1 | - | 1 - 2 | 61 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | formin 1 | FMN1 |
FMN, LD |
-46.8kb | Sense | Chr. 15 - 32765544 - 33194765 on NCBI Build GRCh38 |
| Downstream | uncharacterized LOC101928134 | LOC101928134 | 61.4kb | Sense | Chr. 15 - 33303657 - 33310659 on NCBI Build GRCh38 |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000526835.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000528283.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529623.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000529696.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530020.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | ENST00000530871.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500008962.hg.1 | NR_046005.1 | Refseq |

TMCO5B
Genomic Map