Assay Details
| Gene Symbol: TRIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC029514.1 | - | 1 | 1345 | 92 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 3613 | MIR3613 | -550.0 | Anti-sense | Chr. 13 - 49996415 - 49996501 on NCBI Build GRCh38 | |
| Downstream | potassium channel regulator | KCNRG | DLTET | 16.3kb | Sense | Chr. 13 - 50015254 - 50020922 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TRIM13
Genomic Map