Assay Details
Gene Symbol: LOC284648 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_036490.1 | 1 | 1063 | 63 | ||
GenBank mRNA | BC034591.1 | - | 1 | 949 | 63 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | regulator of G-protein signaling 16 | RGS16 |
A28-RGS14, A28-RGS14P, RGS-R |
-10.8kb | Anti-sense | Chr. 1 - 182598623 - 182604413 on NCBI Build GRCh38 |
Downstream | regulator of G-protein signaling 8 | RGS8 | 26.7kb | Anti-sense | Chr. 1 - 182641814 - 182753291 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001579.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643367 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643355 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643357 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643368 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703024 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643386 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643329 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703005 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643335 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643366 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703016 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703047 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703052 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15149892 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643373 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703013 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703036 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643354 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703011 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703045 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703044 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703040 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643383 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703023 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643384 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643372 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643328 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703035 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703028 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643349 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643371 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643326 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643379 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643385 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703012 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643346 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643341 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703057 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643345 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703034 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643377 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703032 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643347 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20702994 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643361 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703025 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643333 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703014 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15149896 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P20703017 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643387 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00643331 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 705975 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P358115 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 33646 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27284 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P17946116 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P358115 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P358115 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_18_P17946116 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P358115 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT HG-U133+ PM Array Plate | 231427_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR01024386.hg.1 | TC01001579.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01024386.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010850.hg.1 | ENST00000566297.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010850.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010850.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100179710.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100179710.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100179710.hg.1 | TC0100010850.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100179711.hg.1 | ENST00000566297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100179711.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100179711.hg.1 | ENST00000566297.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100179711.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100179711.hg.1 | TC0100010850.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100010850.hg.1 | ENST00000566297.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010850.hg.1 | NR_036490.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010850.hg.1 | NR_036490.1 | Refseq |
Human Genome U133 Plus 2.0 Array | 231427_at | - | - |
Human Genome U133 Plus 2.0 Array | 231427_PM_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 231427_at3 | - | - |
Human Genome U95 Set | 87757_PM_at | - | - |
Human Genome U95 Set | 87757_at | - | - |
Human Genome U133 Set | 231427_PM_at | - | - |
Human Genome U133 Set | 231427_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036231 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A036231 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_3286751 | NR_036490.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_3286751 | BC034591.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1882500 | NR_036490.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1882500 | BC034591.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1882500 | NR_036490.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1882500 | BC034591.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1882500 | NR_036490.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1882500 | BC034591.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 830253 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_83481 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1882500 | NR_036490.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1882500 | BC034591.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1882500 | NR_036490.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1882500 | BC034591.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1882500 | NR_036490.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1882500 | BC034591.1 | human_transcriptome |
