| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011195.hg.1 |
TC06000959.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011195.hg.1 |
ENST00000423208.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06011195.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164210.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164210.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164210.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600164210.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600164210.hg.1 |
TC0600009361.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
ENST00000423208.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600009361.hg.1 |
NR_038906.1 |
Refseq |