Assay Details
| Gene Symbol: DOPEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC036257.1 | - | 4 - 5 | 1519 | 103 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ubiquitin protein ligase E3D | UBE3D |
C6orf157, H10BH, UBE2CBP, YJR141W |
-54.0kb | Sense | Chr. 6 - 82880834 - 83065841 on NCBI Build GRCh38 |
| Downstream | phosphoglucomutase 3 | PGM3 |
AGM1, IMD23, PAGM, PGM 3 |
26.3kb | Sense | Chr. 6 - 83150728 - 83193936 on NCBI Build GRCh38 |
More Information
Set Membership: |
|

DOPEY1
Genomic Map